missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOP1 Recombinant Human Monoclonal Antibody (K94015_6C5), Invitrogen™

Human Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA555995

 View more versions of this product

Catalog No. MA555995


Only null left
Explore available promotions

Description

Description

Product is compatible with indirect ELISA (1 μg/mL, capture antibody). Sequence of this protein is as follows: EHKGPVFAPP YEPLPENVKF YYDGKVMKLS PKAEEVATFF AKMLDHEYTT KEIFRKNFFK DWRKEMTNEE KNIITNLSKC DFTQMSQYFK AQTEARKQMS KEEKLKIKEE NEKLLKEYGF CIMDNHKERI ANFKIEPPGL FRGRGNHPKM GMLKRRIMPE DIIINCSKDA KVPSPPPGHK WKEVRHDNKV TWLVSWTENI

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
Specifications

Specifications

TOP1
Recombinant Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P11387
Top1
Recombinant human TOP1 (immunogen range is 221-420).
200 μg
Primary
Human
Antibody
IgG1
ELISA, Western Blot
K94015_6C5
Unconjugated
Top1
Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; type I DNA topoisomerase
Human
Protein A
RUO
7150
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions