missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOP1 Recombinant Human Monoclonal Antibody (K94015_6C5), Invitrogen™
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555613
Description
Product is compatible with indirect ELISA (1 μg/mL, capture antibody). Sequence of this protein is as follows: EHKGPVFAPP YEPLPENVKF YYDGKVMKLS PKAEEVATFF AKMLDHEYTT KEIFRKNFFK DWRKEMTNEE KNIITNLSKC DFTQMSQYFK AQTEARKQMS KEEKLKIKEE NEKLLKEYGF CIMDNHKERI ANFKIEPPGL FRGRGNHPKM GMLKRRIMPE DIIINCSKDA KVPSPPPGHK WKEVRHDNKV TWLVSWTENI
This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.Specifications
TOP1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P11387 | |
Top1 | |
Recombinant human TOP1 (immunogen range is 221-420). | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
K94015_6C5 | |
Unconjugated | |
Top1 | |
Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; type I DNA topoisomerase | |
Human | |
Protein A | |
RUO | |
7150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |