missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, ACS Reagent Grade, Ricca Chemical
$220.72 - $220.72
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RSOT02004C
|
Ricca Chemical Company
RSOT02004C |
4 L |
Each for $220.72
|
|
|||||
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
CC1=CC=CC=C1 |
C7H8 | |
MFCD00008512 | |
toluene |
Specifications
108-88-3 | |
111°C | |
Amber Glass Bottle | |
C7H8 | |
max. 0.03% | |
CC1=CC=CC=C1 | |
92.14 | |
ACS |
100 | |
4 L | |
max. 0.001% | |
MFCD00008512 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
min. 99.5% | |
Toluene |
Safety and Handling
ShelfLife : 60 months