missing translation for 'onlineSavingsMsg'
Learn More

Tolnaftate 97.0+%, TCI America™

$87.60 - $259.11

Chemical Identifiers

CAS 2398-96-1
Molecular Formula C19H17NOS
Molecular Weight (g/mol) 307.411
MDL Number MFCD00056611
InChI Key FUSNMLFNXJSCDI-UHFFFAOYSA-N
Synonym tolnaftate, tolnaphthate, tinactin, tonoftal, sporiline, tinaderm, tolsanil, aftate, chinofungin, dermoxin
PubChem CID 5510
ChEBI CHEBI:9620
IUPAC Name O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate
SMILES CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T30591G
View Documents
TCI America
T30591G
1 g
Each for $87.60
Only null left
 
T30595G
View Documents
TCI America
T30595G
5 g
Each for $259.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2398-96-1
307.411
FUSNMLFNXJSCDI-UHFFFAOYSA-N
5510
O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate
C19H17NOS
MFCD00056611
tolnaftate, tolnaphthate, tinactin, tonoftal, sporiline, tinaderm, tolsanil, aftate, chinofungin, dermoxin
CHEBI:9620
CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2

Specifications

2398-96-1
White
MFCD00056611
tolnaftate, tolnaphthate, tinactin, tonoftal, sporiline, tinaderm, tolsanil, aftate, chinofungin, dermoxin
CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2
307.411
CHEBI:9620
≥97.0% (HPLC,N)
Tolnaftate
113°C
C19H17NOS
1 g
FUSNMLFNXJSCDI-UHFFFAOYSA-N
O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate
5510
307.41
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1110

RTECSNumber : FD8891300

TSCA : Yes

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'