missing translation for 'onlineSavingsMsg'
Learn More

TOC Standard, According to ISO/CEN EN1484, 500 mg/L±10 mg/L, MilliporeSigma™ Supelco™

$233.23 - $340.99

Specifications

Concentration 500 mg/L ±10 mg/L
Grade Analytical Standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111004584
View Documents
Merck Emd Millipore
76319250MLF
250 mL
Each for $233.23
Only null left
 
111004379
View Documents
Merck Emd Millipore
76319500MLF
500 mL
Each for $340.99
Only null left
 
Description

Description

Specifications

Specifications

500 mg/L ±10 mg/L
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'