Learn More
MilliporeSigma™ Supelco™ TLC Saturation Pads
Pads improve reproducibility and reduce 'edge effects' when performing thin layer chromatography.
$144.92 - $157.16
Specifications
Quantity | 100/Pk. |
---|---|
For Use With (Application) | TLC |
Packing Material | Pk. of 100 Ea. |
Description
Placement of pads with solvent in the development chamber helps to assure rapid and uniform saturation of the atmosphere with solvent vapor.Specifications
100/Pk. | |
TLC | |
Pk. of 100 Ea. |