missing translation for 'onlineSavingsMsg'
Learn More

Titanium Dioxide, USP, 99-100.5%, Spectrum™ Chemical

TI140, 13463-67-7, TiO2

$3154.73 - $8708.17

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
IUPAC Name dioxotitanium
SMILES O=[Ti]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18607426
View Documents
Spectrum Chemical Mfg Cor
TI14012KG
12 kg Poly Pail
Each for $3,154.73
Only null left
 
18607427
View Documents
Spectrum Chemical Mfg Cor
TI14045KGBL
45 kg Fiber Drum
Each for $8,708.17
Only null left
 
Description

Description

Spectrum™ Chemical Titanium Dioxide, USP is used as a food coloring. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

13463-67-7
79.87
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
O=[Ti]=O
O2Ti
MFCD00011269 MFCD00210650
dioxotitanium

Specifications

13463-67-7
12 kg
MFCD00011269 MFCD00210650
O=[Ti]=O
79.87
USP
0.005
1
O2Ti
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
99 to 100.5%
0.005
Poly Pail
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'