missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiglic Acid 98.0+%, TCI America™
$82.02 - $610.67
Chemical Identifiers
CAS | 80-59-1 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00066864 |
InChI Key | UIERETOOQGIECD-ONEGZZNKSA-N |
Synonym | tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid |
PubChem CID | 125468 |
ChEBI | CHEBI:9592 |
IUPAC Name | (2E)-2-methylbut-2-enoic acid |
SMILES | C\C=C(/C)C(O)=O |
Chemical Identifiers
80-59-1 | |
100.12 | |
UIERETOOQGIECD-ONEGZZNKSA-N | |
125468 | |
(2E)-2-methylbut-2-enoic acid |
C5H8O2 | |
MFCD00066864 | |
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid | |
CHEBI:9592 | |
C\C=C(/C)C(O)=O |
Specifications
64°C | |
82°C | |
C5H8O2 | |
3261 | |
UIERETOOQGIECD-ONEGZZNKSA-N | |
(2E)-2-methylbut-2-enoic acid | |
125468 | |
100.12 | |
25 g | |
Tiglic Acid |
80-59-1 | |
White | |
MFCD00066864 | |
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid | |
C\C=C(/C)C(O)=O | |
100.12 | |
CHEBI:9592 | |
≥98.0% (GC,T) | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0609&(2)-0968
RTECSNumber : GQ5430000
TSCA : Yes
Recommended Storage : Refrigerator