missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide TS, 4% w/v, Reagents

Thioacetamide TS, 4% w/v, Meets USP specifications, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F) Manufactured in ISO 9001 facility.

$288.70 - $561.10

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Thiocarbamoyl Hydrazide, Aminothiourea, Hydrazinecarbothioamide
IUPAC Name water
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-333-459
View Documents
Labchem Inc
CT0104001C
1 L
Each for $561.10
Only null left
 
01-333-458
View Documents
Labchem Inc
CT010400500C
500 mL
Each for $288.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Thiocarbamoyl Hydrazide, Aminothiourea, Hydrazinecarbothioamide
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

7732-18-5,62-55-5
100°C
H2O
1.02
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
4%
Liquid
Colorless to Slight Yellow
Glass Bottle
1 L
Thiocarbamoyl Hydrazide, Aminothiourea, Hydrazinecarbothioamide
O
18.02
USP
Thioacetamide TS, 4% w/v
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'