missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamine Hydrochloride, U.S.P. - F.C.C., J.T. Baker™
$8845.29 - $8845.29
Chemical Identifiers
CAS | 67-03-8 |
---|---|
Molecular Formula | C12H18Cl2N4OS |
Molecular Weight (g/mol) | 337.263 |
InChI Key | DPJRMOMPQZCRJU-UHFFFAOYSA-M |
Synonym | thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride |
PubChem CID | 6202 |
ChEBI | CHEBI:49105 |
IUPAC Name | 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride |
SMILES | CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-] |
Chemical Identifiers
67-03-8 | |
337.263 | |
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride | |
CHEBI:49105 | |
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-] |
C12H18Cl2N4OS | |
DPJRMOMPQZCRJU-UHFFFAOYSA-M | |
6202 | |
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride |
Specifications
67-03-8 | |
C12H18Cl2N4OS | |
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride | |
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-] | |
337.263 | |
CHEBI:49105 | |
FCC/USP |
Poly Pail | |
2.5 kg | |
DPJRMOMPQZCRJU-UHFFFAOYSA-M | |
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride | |
6202 | |
337.27 | |
Thiamine Hydrochloride |