Learn More
Thermo Scientific™ Orion™ Ammonium Ion Sensing Module
Get quick, reliable and accurate measurements of dissolved ammonium in aqueous solutions.
Supplier:  Thermo Scientific™ 931801
Catalog No. 13-642-568
Only null left
Explore available promotions
Description
Description
The ammonium module must be used with the 9300BNWP electrode body and double junction reference electrode, Cat. No. 900200.
Warranty period
6 months from date of purchase
Specifications
Specifications
WPBNC
9300BNWP Electrode Body Handle
Ammonia Ion Sensing Module
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories