Learn More
Thermo Scientific™ Labtips Pipette Tips
Produced from the highest quality virgin polypropylene
Supplier: Thermo Scientific™ 94300120
Description
- Double injection molded for maximum sample delivery and best fit for a variety of pipetters
- Available in 200 and 1000μL including bulk and racked formats
Specifications
0.5 to 200 μL | |
Autoclavable | |
LabTip Pipette | |
Polypropylene |
Yellow | |
Bulk | |
LabTip |