missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tft.-45.6 Fixed Angle Rotor
Conduct a variety of applications using the versatile Thermo Scientific™ TFT-45.6 Fixed Angle Rotor, which includes a range of capabilities.
Supplier: Thermo Scientific™ 11637
Description
Conduct a variety of applications using the versatile Thermo Scientific™ TFT-45.6 Fixed Angle Rotor, which includes a range of capabilities.
Compatible with:
Thermo Scientific Sorvall WX Ultra Series Floor Model Centrifuge
Recommended for:
Serum lipoprotein flotation, multiplesample DNA and virus isolation.
Specifications
TFT-45.6 Fixed Angle Rotor | |
Fixed Angle Rotor | |
238,859 x g (Outer Row), 204,252 x g (Inner Row) | |
105.6 mm Outer, 90.3 mm Inner | |
44.1 Outer Row/49.6 Inner Row | |
13 mm | |
Thermo Scientific™ Sorvall™ WX ultraCentrifuges | |
205,383 x g Outer/170,755 x g Inner |
Titanium | |
40 x 6 mL | |
45,000 rpm | |
76.0 mm Outer, 60.7 mm Inner | |
66 mm | |
24.3 lb. (10.9 kg) | |
20° | |
171,906 x g Outer; 137,299 x g Inner |