missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ Isolator Cleaning Tool Replacement Handles

$82.03 - $87.96

Specifications

Product Type Replacement Handle
Color White
Autoclavable Autoclavable
For Use With (Equipment) Isolator Cleaning Tool
Material Fiberglass
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Handle Price Quantity  
Catalog Number Mfr. No. Length (English) Handle Price Quantity  
19-222-370
View Documents
Texwipe™
TX7120
12 in.
Case for $87.96
Only null left
 
19-222-371
View Documents
Texwipe™
TX7121
18 in.
Each for $82.03
Only null left
 
Description

Description

Specifications

Specifications

Replacement Handle
Autoclavable
Fiberglass
White
Isolator Cleaning Tool
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'