missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium chloride, For ion pair chromatography (AT), ≥99.0% (AT), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 74202250GF
Specifications
>300°C | |
250 g | |
MFCD00011628 | |
109.60 | |
Ion Pair Chromatography |
75-57-0 | |
(CH3)4 N(Cl) | |
TMA | |
≥99.0% (AT) |
Chemical Identifiers
MFCD00011628 | |
TMA |
Safety and Handling
RTECSNumber : BS7700000