missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Stabilized with 0.025% BHT, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T10254LTGL

 View more versions of this product

Catalog No. 18614362


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

109-99-9
4 L
Amber Glass Bottle
0.0005
ACS
1
To Pass Test
0.0003
0.99

Chemical Identifiers

109-99-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.