missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, contains no preservative, >99.8%, Honeywell™

$2514.25 - $2817.55

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
6001680
missing translation for 'viewDocuments'
Honeywell Inc
AH3404
4 x 4 L Glass Bottle
Case of 4 Each for $2,514.25
Only null left
 
6001681
missing translation for 'viewDocuments'
Honeywell Chemicals
AH3404PC
4 x 4 L Poly-coated Bottle
Case of 4 Each for $2,817.55
Only null left
 
Description

Description

Specifications

Chemical Identifiers

109-99-9
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1

Specifications

109-99-9
100.0000%
66°C
−14°C
C4H8O
UN2056
102391
Miscible in Water at 20°C
C1CCOC1
72.11
CHEBI:26911
>99.8%
Tetrahydrofuran, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, contains no preservative, >99.8%
0.18 AU max. at 250 nm, 0.02 AU max. at 300 nm
0.8892 g/cm3
4 x 4 L
Glass Bottle
MFCD00005356
0.03% max.
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
HPLC
Videos
Safety and Handling

Safety and Handling

ShelfLife : 365 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran

EINECSNumber : 203-726-8

SDS
Documents

Documents