missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluorotetracyanoquinodimethane (purified by sublimation) 98.0+%, TCI America™

$454.83 - $2046.73

Chemical Identifiers

CAS 29261-33-4
Molecular Formula C12F4N4
Molecular Weight (g/mol) 276.154
MDL Number MFCD00042382
InChI Key IXHWGNYCZPISET-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 2,2'-perfluorocyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tcnqf4, 2,3,5,6-tetrafluorotetracyanoquinodimethane, f4-tcnq, tetrafluorotetracyanoquinodimethane, 2-4-dicyanomethylidene-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene propanedinitrile, f4tcnq, tetrafluorotetracyanoquinodimethane anion radical, tetrafluorotetracyanoquinodimethane purifiedbysubl
PubChem CID 2733307
IUPAC Name 2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES C(#N)C(=C1C(=C(C(=C(C#N)C#N)C(=C1F)F)F)F)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T1131100MG
View Documents
TCI America
T1131100MG
100 mg
Each for $454.83
Only null left
 
T11311G
View Documents
TCI America
T11311G
1 g
Each for $2,046.73
Only null left
 
Description

Description

Specifications

Chemical Identifiers

29261-33-4
276.154
IXHWGNYCZPISET-UHFFFAOYSA-N
2733307
C(#N)C(=C1C(=C(C(=C(C#N)C#N)C(=C1F)F)F)F)C#N
C12F4N4
MFCD00042382
2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 2,2'-perfluorocyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tcnqf4, 2,3,5,6-tetrafluorotetracyanoquinodimethane, f4-tcnq, tetrafluorotetracyanoquinodimethane, 2-4-dicyanomethylidene-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene propanedinitrile, f4tcnq, tetrafluorotetracyanoquinodimethane anion radical, tetrafluorotetracyanoquinodimethane purifiedbysubl
2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile

Specifications

29261-33-4
C12F4N4
100 mg
2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 2,2'-perfluorocyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tcnqf4, 2,3,5,6-tetrafluorotetracyanoquinodimethane, f4-tcnq, tetrafluorotetracyanoquinodimethane, 2-4-dicyanomethylidene-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene propanedinitrile, f4tcnq, tetrafluorotetracyanoquinodimethane anion radical, tetrafluorotetracyanoquinodimethane purifiedbysubl
C(#N)C(=C1C(=C(C(=C(C#N)C#N)C(=C1F)F)F)F)C#N
276.154
276.15
Crystalline Powder
300°C
MFCD00042382
3439
IXHWGNYCZPISET-UHFFFAOYSA-N
2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
2733307
≥98.0% (N)
Tetrafluorotetracyanoquinodimethane (purified by sublimation) [Organic Electronic Material]
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'