missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylthiuram Disulfide 97.0+%, TCI America™
$33.56 - $113.71
Chemical Identifiers
CAS | 97-77-8 |
---|---|
Molecular Formula | C10H20N2S4 |
Molecular Weight (g/mol) | 296.524 |
MDL Number | MFCD00009048 |
InChI Key | AUZONCFQVSMFAP-UHFFFAOYSA-N |
Synonym | disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd |
PubChem CID | 3117 |
ChEBI | CHEBI:4659 |
IUPAC Name | diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate |
SMILES | CCN(CC)C(=S)SSC(=S)N(CC)CC |
Chemical Identifiers
97-77-8 | |
296.524 | |
AUZONCFQVSMFAP-UHFFFAOYSA-N | |
3117 | |
diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate |
C10H20N2S4 | |
MFCD00009048 | |
disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
CHEBI:4659 | |
CCN(CC)C(=S)SSC(=S)N(CC)CC |
Specifications
97-77-8 | |
White-Yellow | |
MFCD00009048 | |
3077 | |
AUZONCFQVSMFAP-UHFFFAOYSA-N | |
diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate | |
3117 | |
296.52 | |
Crystalline Powder |
70°C | |
C10H20N2S4 | |
25 g | |
disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
CCN(CC)C(=S)SSC(=S)N(CC)CC | |
296.524 | |
CHEBI:4659 | |
≥97.0% (T) | |
Tetraethylthiuram Disulfide |
Safety and Handling
EINECSNumber : (2)-1820
RTECSNumber : JO1225000
TSCA : Yes