missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecane, (n-Tetradecane), Reagents
Tetradecane, (n-Tetradecane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$488.57 - $2,182.83
Chemical Identifiers
CAS | 629-59-4 |
---|---|
Molecular Formula | C14H30 |
Molecular Weight (g/mol) | 198.39 |
InChI Key | BGHCVCJVXZWKCC-UHFFFAOYSA-N |
Synonym | n-Tetradecane, Methyl tridecane |
IUPAC Name | tetradecane |
SMILES | CCCCCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-336-934
|
Labchem Inc
C2337530100C |
100 mL |
N/A
|
|
|||||
01-336-933
|
Labchem Inc
C2337530450C |
450 mL |
N/A
|
|
|||||
Chemical Identifiers
629-59-4 | |
198.39 | |
n-Tetradecane, Methyl tridecane | |
CCCCCCCCCCCCCC |
C14H30 | |
BGHCVCJVXZWKCC-UHFFFAOYSA-N | |
tetradecane |
Specifications
629-59-4 | |
4°C | |
253°C | |
Glass Bottle | |
100 mL | |
n-Tetradecane, Methyl tridecane | |
CCCCCCCCCCCCCC | |
198.39 | |
Liquid |
253°C | |
Colorless | |
4°C | |
C14H30 | |
0.763 | |
BGHCVCJVXZWKCC-UHFFFAOYSA-N | |
tetradecane | |
Reagent | |
Tetradecane, (n-Tetradecane) |