missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(tert-Butyldimethylsilyloxy)malononitrile 93.0+%, TCI America™
Supplier: TCI America B53701G
Specifications
(tert-Butyldimethylsilyloxy)malononitrile | |
Yellow | |
MFCD27942977 | |
3276 | |
AIQDCGCDLHCZHB-UHFFFAOYSA-N | |
2-[tert-butyl(dimethyl)silyl]oxypropanedinitrile | |
10856410 | |
≥93.0% (GC) |
128302-78-3 | |
C9H16N2OSi | |
1 g | |
(tert-Butyldimethylsilyloxy)propanedinitrile, MAC-TBS Reagent | |
CC(C)(C)[Si](C)(C)OC(C#N)C#N | |
196.325 | |
196.33 | |
Liquid |
Chemical Identifiers
128302-78-3 | |
196.325 | |
AIQDCGCDLHCZHB-UHFFFAOYSA-N | |
10856410 | |
CC(C)(C)[Si](C)(C)OC(C#N)C#N |
C9H16N2OSi | |
MFCD27942977 | |
(tert-Butyldimethylsilyloxy)propanedinitrile, MAC-TBS Reagent | |
2-[tert-butyl(dimethyl)silyl]oxypropanedinitrile |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator