missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 2,2,2-Trichloroacetimidate 95.0+%, TCI America™
$109.60 - $332.60
Chemical Identifiers
CAS | 98946-18-0 |
---|---|
Molecular Formula | C6H10Cl3NO |
Molecular Weight (g/mol) | 218.50 |
MDL Number | MFCD00077410 |
InChI Key | CQXDYHPBXDZWBA-UHFFFAOYSA-N |
Synonym | tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine |
PubChem CID | 2734700 |
IUPAC Name | tert-butyl 2,2,2-trichloroethanimidate |
SMILES | CC(C)(C)OC(=N)C(Cl)(Cl)Cl |
Chemical Identifiers
98946-18-0 | |
218.50 | |
CQXDYHPBXDZWBA-UHFFFAOYSA-N | |
2734700 | |
CC(C)(C)OC(=N)C(Cl)(Cl)Cl |
C6H10Cl3NO | |
MFCD00077410 | |
tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine | |
tert-butyl 2,2,2-trichloroethanimidate |
Specifications
98946-18-0 | |
69°C | |
MFCD00077410 | |
1993 | |
CQXDYHPBXDZWBA-UHFFFAOYSA-N | |
tert-butyl 2,2,2-trichloroethanimidate | |
2734700 | |
≥95.0% (GC) | |
Liquid |
Yellow | |
C6H10Cl3NO | |
5 mL | |
tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine | |
CC(C)(C)OC(=N)C(Cl)(Cl)Cl | |
218.50 | |
218.50 | |
Reagent | |
tert-Butyl 2,2,2-Trichloroacetimidate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator