missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™

CHROMASOLV™, for HPLC,≥99.5%

Supplier:  Honeywell-Chromasolv 3082504X25L

 View more versions of this product

Catalog No. 6001642


Only null left
Explore available promotions

Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Specifications

tert-Butanol
CHROMASOLV™, for HPLC, ≥99.5%
max. 0.01 at 350 nm, max. 0.01 at 300 nm, max. 0.20 at 250 nm, max. 0.50 at 230 nm, max. 1.00 at 215 nm
0.775 g/mL at 25 °C(lit.)
4 x 2.5 L
Glass Bottle
(CH3)3COH
UN1120
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CC(C)(C)O
74.123
CHEBI:45895
>99.5%
23°C to 26°C (literature)
75-65-0
100.0000%
81°C to 83°C
11°C
C4H10O
MFCD00004464
906698
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12
HPLC

Chemical Identifiers

75-65-0
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol
C4H10O
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CHEBI:45895
CC(C)(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 720 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Butanols

EINECSNumber : 200-889-7

Recommended Storage : Room Temp

SDS
Documents

Documents

Promotions

Promotions