Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Polyurethane Coating
Knit with high-tenacity composite yarns designed to meet a range of applications. Polyurethane coating offers a high coefficient of friction that is non-shedding.
$11.12 - $12.52
Specifications
Coating Material | Polyurethane |
---|---|
Color | Gray |
Cut Resistance Rating | ANSI A2 |
Material | Composite Fiber |
Product Type | Cut-resistant Gloves |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
29-636-3135
|
Superior Glove™
S13TAGPU6 |
6 |
Pair for $12.34
Case of 240 Pair for $2,628.00
|
|
|||||
29-636-3136
|
Superior Glove™
S13TAGPU7 |
7 |
Pair for $12.52
Case of 240 Pair for $2,666.40
|
|
|||||
29-636-3137
|
Superior Glove™
S13TAGPU8 |
8 |
Pair for $12.18
Case of 240 Pair for $2,594.40
|
|
|||||
29-636-3139
|
Superior Glove™
S13TAGPU9 |
9 |
Pair for $12.10
Case of 240 Pair for $2,577.60
|
|
|||||
29-636-3138
|
Superior Glove™
S13TAGPU10 |
10 |
Pair for $11.99
Case of 240 Pair for $2,553.60
|
|
|||||
29-636-3140
|
Superior Glove™
S13TAGPU11 |
11 |
Pair for $12.18
Case of 240 Pair for $2,594.40
|
|
|||||
17-401-254
|
Superior Glove™
S13TAGPU12 |
12 |
Pair for $11.12
Case of 240 Pair for $2,368.80
|
|
|||||
Description
- Continuous filament yarn is stronger than steel on an equal-weight basis
- 1250 grams of cut protection
- Lint-free
Withstands oils and solvents
Specifications
Polyurethane | |
ANSI A2 | |
Cut-resistant Gloves | |
Knit |
Gray | |
Composite Fiber | |
Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping |