missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chiron Telomeric Alcohol Standards
PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.
$722.87 - $932.60
Specifications
Concentration | 50 μg/mL |
---|---|
Type | Reference Material |
Solvent or Matrix | Methanol |
Quantity | 1 mL |
Catalog Number | Mfr. No. | Chemical Name or Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Chemical Name or Material | Price | Quantity | |||||
28-449-50ME
|
Chiron
2844950ME |
1H,1H,2H,2H,3H,3H-Perfluorononan-1-ol |
Each for $932.60
|
|
|||||
28-479-50ME
|
Chiron
2847950ME |
1H,1H,2H,2H-Perfluoro-7-methyloctan-1-ol |
Each for $722.87
|
|
|||||
Description
- Various singles available
- Solutions in assorted solvents
- Useful for various matrices
- Custom standards of other PFAS compounds available on demand
Specifications
50 μg/mL | |
Reference Material | |
Methanol | |
1 mL |