missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tartaric Acid, Purified, Reagents
Tartaric Acid, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2116500500A5
Specifications
Tartaric Acid, Purified | |
87-69-4 | |
Colorless/White | |
MFCD00064207 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
2,3-dihydroxybutanedioic acid | |
Pure | |
Poly Bottle | |
1.7598 |
168°C to 170°C | |
275°C | |
C4H6O6 | |
DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid | |
OC(C(O)C(O)=O)C(O)=O | |
150.09 | |
500 g | |
Solid |
Chemical Identifiers
87-69-4 | |
150.09 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
2,3-dihydroxybutanedioic acid |
C4H6O6 | |
MFCD00064207 | |
DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid | |
OC(C(O)C(O)=O)C(O)=O |