missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T101525KG

 View more versions of this product

Catalog No. 18614315


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

87-69-4
99%
2.5 kg
0.0002
1
ACS
Poly Bottle

Chemical Identifiers

87-69-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.