missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tartaric Acid, Certified, 10.0% (w/v) ±0.5% (w/v), LabChem™
$110.37 - $161.80
Chemical Identifiers
CAS | 87-69-4 |
---|---|
Molecular Formula | C4H6O6 |
Molecular Weight (g/mol) | 150.09 |
MDL Number | MFCD00064207 |
InChI Key | FEWJPZIEWOKRBE-UHFFFAOYNA-N |
Synonym | l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid |
PubChem CID | 444305 |
ChEBI | CHEBI:15671 |
IUPAC Name | 2,3-dihydroxybutanedioic acid |
SMILES | OC(C(O)C(O)=O)C(O)=O |
Chemical Identifiers
87-69-4 | |
150.09 | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
444305 | |
2,3-dihydroxybutanedioic acid |
C4H6O6 | |
MFCD00064207 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
CHEBI:15671 | |
OC(C(O)C(O)=O)C(O)=O |
Specifications
87-69-4,7732-18-5 | |
Colorless to Yellow | |
C4H6O6 | |
l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
2,3-dihydroxybutanedioic acid | |
444305 | |
150.09 | |
Certified | |
500 mL | |
Liquid | |
Traceable to NIST |
90.4,9.6 | |
C4H6O6 | |
MFCD00064207 | |
Soluble in water | |
OC(C(O)C(O)=O)C(O)=O | |
150.09 | |
CHEBI:15671 | |
10.0% ±0.5% (w/v) | |
1.04g/mL | |
Poly Bottle | |
1.04g/mL | |
Tartaric Acid |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Danger
Recommended Storage : Refrigerate