missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, ACS Reagent, Reagents

Tartaric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

$277.37 - $832.20

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-338-157
View Documents
Labchem Inc
C1116500125D2
125 g
Each for $277.37
Only null left
 
01-338-156
View Documents
Labchem Inc
C1116500500A5
500 g
Each for $832.20
Only null left
 
Description

Description

Specifications

Chemical Identifiers

87-69-4
150.09
FEWJPZIEWOKRBE-UHFFFAOYNA-N
2,3-dihydroxybutanedioic acid
C4H6O6
MFCD00064207
DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid
OC(C(O)C(O)=O)C(O)=O

Specifications

168°C to 170°C
275°C
C4H6O6
DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid
OC(C(O)C(O)=O)C(O)=O
150.09
125 g
Solid
Tartaric Acid, ACS Reagent
87-69-4
Colorless/White
MFCD00064207
FEWJPZIEWOKRBE-UHFFFAOYNA-N
2,3-dihydroxybutanedioic acid
ACS
Glass Bottle
1.7598
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'