Learn More
Gibco™ TAP Growth Media, optimized for Chlamydomonas culture DFS Item
TAP Growth Media, optimized for Chlamydomonas culture
$700.00 - $700.00
Specifications
Form Liquid
Product Type Growth Medium
Preparation Method None (Ready-to-Use)
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A1379802 DFS Item
View Documents
Gibco™
A1379802
6 x 1 L N/A
Only null left
Description
Description
Gibco™ TAP Media is optimized for the growth and maintenance of Chlamydomonas reinhardtii. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.
Each Gibco™ TAP Media comes in the award winning Gibco™ bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helping you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Chlamydomonas culture, results in better overall efficiency and more robust data (Fig. 1, 2).
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Specifications
Liquid
Growth Medium
None (Ready-to-Use)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'