missing translation for 'onlineSavingsMsg'
Learn More

Tantalum tubing, 3.18mm (0.125in) OD, 2.26mm (0.089in) ID, 99.9% (metals basis), Thermo Scientific™

$1090.96 - $1090.96

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44923BW
View Documents
Thermo Scientific Chemicals
44923BW
50 cm
Each for $1,090.96
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]

Specifications

7440-25-7
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
CHEBI:33348
Odorless
(metals basis)
Ta
50 cm
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
99.9%
Tantalum tubing, OD: 3.18mm (0.125 in.), ID: 2.26mm (0.089 in.)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-135-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents