missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin 1 Monoclonal Antibody (K1E005_2F9), Invitrogen™
Mouse Monoclonal Antibody
$1693.48 - $2847.38
Specifications
Antigen | Synaptotagmin 1 |
---|---|
Clone | K1E005_2F9 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Description
Sequence of this protein is as follows: KKGKEKGGKN AINMKDVKDL GKTMKDQALK DDDAETGLTD GEEKEEPKEE EKLGKLQYSL DYDFQNNQLL VGIIQAAELP ALDMGGTSDP YVKVFLLPDK KKKFETKVHR KTLNPVFNEQ FTFKVPYSEL GGKTLVMAVY DFDRFSKHDI IGEFKVPMNT VDFGHVTEEW RDLQSAEKEE QEKLGDICFS LRYVPTAGKL TVVILEAKNL KKMDVGGLSD PYVKIHLMQN GKRLKKKKTT IKKNTLNPYY NESFSFEVPF EQIQKVQVVV TVLDYDKIGK NDAIGKVFVG YNSTGAELRH WSDMLANPRR PIAQWHTLQV EEEVDAMLAV
Synaptotagmin 1 Monoclonal antibody specifically detects Synaptotagmin 1 in Human, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western BlotSpecifications
Synaptotagmin 1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide | |
AW124717; DKFZp781D2042; FLJ42519; G630098F17Rik; P65; SV DKFZp781D2042; SV SYT; svp65; Syb1; synaptotagmin 1; synaptotagmin 1 L homeolog; synaptotagmin 2; synaptotagmin I; synaptotagmin I protein; synaptotagmin I variant b; Synaptotagmin II; Synaptotagmin XI; synaptotagmin-1; Synaptotagmin-2; SYT; Syt1; syt1.L; SYT2; SytI; SytII; XELAEV_18022019mg | |
Recombinant human SYT1 (immunogen range is 91-420). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Syt1 |
K1E005_2F9 | |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
P21579, P21707 | |
25716, 6857 | |
IgG1 | |
Protein A | |
Antibody |