missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Certified, 0.0200N ±0.0002N (0.01M), LabChem™
$37.40 - $262.33
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
LC256601
|
LabChem
LC256601 |
500 mL |
Each for $37.40
|
|
|||||
LC256602
|
LabChem
LC256602 |
1 L |
Each for $43.80
|
|
|||||
LC256604
|
LabChem
LC256604 |
4 L |
Each for $94.17
|
|
|||||
LC256606
|
LabChem
LC256606 |
10 L |
Each for $187.70
|
|
|||||
LC256605
|
LabChem
LC256605 |
20 L |
Each for $262.33
|
|
|||||
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
Colorless | |
0.0200N ±0.0002N (0.01M) | |
99.9,0.1 | |
Poly Bottle | |
Traceable to NIST | |
500 mL | |
H2O4S | |
H2SO4 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
Sulfuric Acid |
7664-93-9,7732-18-5 | |
1g/mL | |
Passes Test | |
1g/mL | |
Sulfur compounds | |
1cSt | |
Liquid | |
MFCD00064589 | |
Soluble in water | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
Certified |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Recommended Storage : Room Temperature