missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, ACS Reagent, Reagents
Sulfuric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$98.63 - $381.73
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-336-807
|
Labchem Inc
C51026401A |
1 L |
Each for $123.23
|
|
|||||
01-336-806
|
Labchem Inc
C51026404A |
4 L |
Each for $381.73
|
|
|||||
01-336-805
|
Labchem Inc
C5102640500A |
500 mL |
Each for $98.63
|
|
|||||
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate | |
OS(O)(=O)=O |
Specifications
0 | |
7664-93-9,7732-18-5 | |
Poly Bottle | |
1 L | |
Liquid | |
Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate | |
OS(O)(=O)=O | |
98.07 | |
Sulfuric Acid, ACS Reagent |
Colorless | |
330°C | |
1.84 | |
H2O4S | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
ACS |