missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.417 Normal, Reagents
Sulfuric Acid, 0.417 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
$356.57 - $519.67
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Sulphuric Acid, Dihydrogen Sulfate |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-333-539
|
Labchem Inc
CS20130010F |
10 L |
Each for $356.57
|
|
|||||
01-333-538
|
Labchem Inc
CS20130020F |
20 L |
Each for $519.67
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Sulphuric Acid, Dihydrogen Sulfate | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
<1 | |
7732-18-5,7664-93-9 | |
Cubitainer | |
10 L | |
Liquid | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent |
Colorless | |
100°C | |
1.0136 | |
H2O | |
Sulphuric Acid, Dihydrogen Sulfate | |
O | |
18.02 | |
Sulfuric Acid, 0.417 Normal |