missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.02N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$730.76 - $990.50
Chemical Identifiers
CAS | 7664-93-9 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
7664-93-9 | |
1L = 1.02kg | |
H2O4S | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
BAKER ANALYZED™ Reagent |
Narrow Mouth Polyethylene Bottle | |
1 L | |
MFCD00064589 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
Sulfuric Acid, 0.02N Volumetric Solution |