missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Lump, Spectrum™ Chemical

$262.32 - $5885.56

Chemical Identifiers

CAS 7704-34-9
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614203
View Documents
Spectrum Chemical Mfg Cor
S1725500GM
500 g Poly Bottle
Each for $262.32
Only null left
 
18614201
View Documents
Spectrum Chemical Mfg Cor
S1725100LBBL
100 lb. Fiber Drum
Each for $5,885.56
Only null left
 
18614202
View Documents
Spectrum Chemical Mfg Cor
S172525KG
2.5 kg Poly Bottle
Each for $748.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7704-34-9

Specifications

7704-34-9
500 g
Poly Bottle
1
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.