missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic Acid, ACS Reagent, Reagents
Sulfamic Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C211430025A5
Specifications
Sulfamic Acid, ACS Reagent | |
205°C | |
Poly Bottle | |
2.5 kg | |
Solid | |
IIACRCGMVDHOTQ-UHFFFAOYSA-N | |
sulfamic acid | |
ACS |
White to Off-White | |
5329-14-6 | |
2.15 | |
H3NO3S | |
Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid | |
NS(O)(=O)=O | |
97.09 |
Chemical Identifiers
5329-14-6 | |
97.09 | |
Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid | |
NS(O)(=O)=O |
H3NO3S | |
IIACRCGMVDHOTQ-UHFFFAOYSA-N | |
sulfamic acid |