missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic Acid, 10% w/v, Reagents
Sulfamic Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$150.97 - $395.27
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-333-520
|
Labchem Inc
CS21000010F |
10 L |
Each for $265.07
|
|
|||||
01-333-519
|
Labchem Inc
CS21000020F |
20 L |
Each for $395.27
|
|
|||||
01-333-518
|
Labchem Inc
CS2100004A |
4 L |
Each for $150.97
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
100°C | |
1.05 | |
H2O | |
Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid | |
O | |
18.02 | |
Reagent |
7732-18-5,5329-14-6 | |
Cubitainer | |
10 L | |
Liquid | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
10% w/v | |
Sulfamic Acid, 10% w/v |