missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadoxine, 98%
CAS: 2447-57-6 | C12H14N4O4S | 310.33 g/mol
$79.08 - $829.70
Chemical Identifiers
CAS | 2447-57-6 |
---|---|
Molecular Formula | C12H14N4O4S |
Molecular Weight (g/mol) | 310.33 |
MDL Number | MFCD00792890 |
InChI Key | PJSFRIWCGOHTNF-UHFFFAOYSA-N |
Synonym | sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil |
PubChem CID | 17134 |
ChEBI | CHEBI:9329 |
IUPAC Name | 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide |
SMILES | COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AAJ6626806
|
Thermo Scientific Chemicals
J6626806 |
5 g | Glass bottle |
Each for $79.08
|
|
||||
AAJ6626814
|
Thermo Scientific Chemicals
J6626814 |
25 g | Plastic bottle |
Each for $254.21
|
|
||||
AAJ6626822
|
Thermo Scientific Chemicals
J6626822 |
100 g | Plastic bottle |
Each for $829.70
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2447-57-6 | |
310.33 | |
PJSFRIWCGOHTNF-UHFFFAOYSA-N | |
17134 | |
4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide |
C12H14N4O4S | |
MFCD00792890 | |
sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil | |
CHEBI:9329 | |
COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC |
Specifications
2447-57-6 | |
C12H14N4O4S | |
5 g | |
sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil | |
PJSFRIWCGOHTNF-UHFFFAOYSA-N | |
4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide | |
17134 | |
310.33 | |
Powder |
Glass bottle | |
MFCD00792890 | |
625453 | |
Soluble in ethanol & ammonium hydroxide (9:1) at 20mg/ml | |
COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC | |
310.33 | |
CHEBI:9329 | |
98% | |
Sulfadoxine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-504-9
RTECSNumber : DA9500000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only