missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic Anhydride 95.0+%, TCI America™
$35.41 - $54.06
Chemical Identifiers
CAS | 108-30-5 |
---|---|
Molecular Formula | C4H4O3 |
Molecular Weight (g/mol) | 100.07 |
MDL Number | MFCD00005525 |
InChI Key | RINCXYDBBGOEEQ-UHFFFAOYSA-N |
Synonym | succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa |
PubChem CID | 7922 |
ChEBI | CHEBI:36595 |
IUPAC Name | oxolane-2,5-dione |
SMILES | O=C1CCC(=O)O1 |
Chemical Identifiers
108-30-5 | |
100.07 | |
RINCXYDBBGOEEQ-UHFFFAOYSA-N | |
7922 | |
oxolane-2,5-dione |
C4H4O3 | |
MFCD00005525 | |
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa | |
CHEBI:36595 | |
O=C1CCC(=O)O1 |
Specifications
120°C | |
131°C | |
C4H4O3 | |
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa | |
O=C1CCC(=O)O1 | |
100.07 | |
CHEBI:36595 | |
≥95.0% (T) | |
Pellets |
108-30-5 | |
White | |
MFCD00005525 | |
RINCXYDBBGOEEQ-UHFFFAOYSA-N | |
oxolane-2,5-dione | |
7922 | |
100.07 | |
25 g | |
Succinic Anhydride |
Safety and Handling
EINECSNumber : (2)-0921
RTECSNumber : WN0875000
TSCA : Yes