missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid 99.0+%, TCI America™

$37.29 - $67.11

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S010025G
View Documents
TCI America
S010025G
25 g
Each for $37.29
Only null left
 
S0100500G
View Documents
TCI America
S0100500G
500 g
Each for $67.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-15-6
118.09
KDYFGRWQOYBRFD-UHFFFAOYSA-N
1110
butanedioic acid
C4H6O4
MFCD00002789
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
CHEBI:15741
OC(=O)CCC(O)=O

Specifications

2.7 (0.1M H2O soln.)
110-15-6
White
MFCD00002789
KDYFGRWQOYBRFD-UHFFFAOYSA-N
butanedioic acid
1110
118.09
25 g
Succinic Acid
187°C
235°C
C4H6O4
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
OC(=O)CCC(O)=O
118.09
CHEBI:15741
≥99.0% (T)
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0846

RTECSNumber : WM4900000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'