missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
S1685, 110-15-6, C4H6O4
$222.40 - $543.47
Chemical Identifiers
CAS | 110-15-6 |
---|---|
Molecular Formula | C4H6O4 |
Molecular Weight (g/mol) | 118.09 |
MDL Number | MFCD00002789 |
InChI Key | KDYFGRWQOYBRFD-UHFFFAOYSA-N |
IUPAC Name | butanedioic acid |
SMILES | OC(=O)CCC(O)=O |
Description
Spectrum™ Chemical Succinic Acid, Crystal, Reagent, ACS is a white, odorless solid that is a dicarboxylic acid. Advances in industrial biotechnology that seek to displace petroleum-based chemicals in industrial applications are fueling growth in this product. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
110-15-6 | |
118.09 | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
OC(=O)CCC(O)=O |
C4H6O4 | |
MFCD00002789 | |
butanedioic acid |
Specifications
185°C to 191°C | |
1 | |
MFCD00002789 | |
OC(=O)CCC(O)=O | |
118.09 | |
ACS | |
Poly Bottle |
110-15-6 | |
C4H6O4 | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
butanedioic acid | |
99% | |
125 g | |
0.0002 |