missing translation for 'onlineSavingsMsg'
Learn More

Strychnine Hydrochloride, Dihydrate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor ST145100GM

 View more versions of this product

Catalog No. 18614117


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

6101-04-8
100 g
Reagent
1
98%
Amber Glass Bottle

Chemical Identifiers

6101-04-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food, drug or pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.