missing translation for 'onlineSavingsMsg'
Learn More

Strychnine, Free Base, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor ST14225GM

 View more versions of this product

Catalog No. 18614123


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

57-24-9
25 g
Reagent
1
98%
Amber Glass Bottle

Chemical Identifiers

57-24-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food, drug or pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.