missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Strontium ranelate

Chemical Identifiers

CAS 135459-87-9
Chemical Name or Material Strontium ranelate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC463010050
View Documents
Thermo Scientific™
463010050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC463010250
View Documents
Thermo Scientific™
463010250
25 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC463011000
View Documents
Thermo Scientific™
463011000
100 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

135459-87-9
Strontium ranelate

Specifications

135459-87-9
5 g
Strontium ranelate
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye irritation.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only