missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptozotocin, MP Biomedicals™
Used in the treatment of pancreatic islet cell tumors. Streptozotocin, MP Biomedicals is a glucosamine-nitrosourea, a DNA-alkylating agent that enters cells via the GLUT2 glucose transport protein. It is used to treat malignant insulinoma and to induce Ty
$751.87 - $751.87
Chemical Identifiers
CAS | 18883-66-4 |
---|---|
Molecular Formula | C8H15N3O7 |
Molecular Weight (g/mol) | 265.222 |
InChI Key | ZSJLQEPLLKMAKR-UHEPRFFZSA-N |
Synonym | N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozocin, STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose |
PubChem CID | 45357367 |
IUPAC Name | 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea |
SMILES | CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
MP21005575
|
MP Biomedicals Inc
021005575 |
500 mg |
Each for $751.87
|
|
|||||
Description
Streptozocin is used to treat pancreatic islet cell tumors, including malignant insulinoma. It is also used to induce Type 1 diabetes in animals for medical research purposes.
- An N-nitroso-containing compound that acts as a nitric oxide donor in pancreatic islets
- Induces death of insulin-secreting cells, producing an animal model of diabetes
- Potent DNA methylating agent that induces chromosomal breakage
- Cytotoxic to neuroendocrine tumor cell lines that express the GLUT2 glucose transporter
Chemical Identifiers
18883-66-4 | |
265.222 | |
N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozocin, STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose | |
1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea |
C8H15N3O7 | |
ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
45357367 | |
CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O |
Specifications
Powder | |
121°C (decomposition) (literature) | |
500 mg | |
N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozocin, STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose | |
ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea | |
45357367 | |
∼75% α-anomer basis, ≥97% |
18883-66-4 | |
EmM (228nm): 6.36 (ethanol) | |
C8H15N3O7 | |
Soluble in water (50mg/mL, clear to slightly hazy, light yellow solution) and ethanol. | |
CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O | |
265.222 | |
265.22 |
Safety and Handling
Recommended Storage : −20°C, desiccate, store under nitrogen