missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ STP 120 Spin Tissue Processor

Process biological specimens from chemical fixation to paraffin infiltration with the Epredia™ STP 120 Spin Tissue Processor.

Supplier:  Epredia™ 813150

 View more versions of this product

Catalog No. 36-101-3476

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Immediate and delayed start processing modes
  • Programmable spinning speed of either 60 or 70 rpm; spinning can also be programmed off
  • Programmable immersion time in each station (from 1 minute to 99 hours: 59 minutes)
  • Basket capacity of 120 or 240 cassettes
  • Battery backup system in case of power failure

Performance:

  • Reagent vessel tops and charcoal-enhanced ventilation help control processing vapors
  • Up to 10 different processing programs maintained
  • Cassette baskets spin counterclockwise and clockwise within reagent container to improve processing
  • Reagent carryover is reduced through gentle centrifugal spinning of basket above reagent vessel
  • Each reagent vessel holds 1.8 L of processing reagent
  • Physical Specifications:

  • Electrical Requirements: 100-240 V, 50/60 Hz
  • Diameter: 85 cm (33.5 in.)
  • Height: 50 cm opening to 70 cm (19.6 in. opening to 27.5 in.)
  • Weight: 154 lb. (70 kg)
  • Includes:

  • Basic instrument
  • Instruction manual
  • Ten reagent vessels and two paraffin baths for single basket operation (120-cassette capacity) or nine reagent vessels and three paraffin baths for double basket operation (240-cassette capacity)
  • Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions