missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ SterilElite™ Vertical Autoclave
Vertical advanced autoclave includes vacuum and dry function
$25692.86 - $42218.80
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Quality Management System |
For Use With (Application) | Decontamination of Biohazardous Waste |
Product Type | Autoclave |
Temperature (English) | 249°F to 275°F |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
STE50VAR
|
Fisherbrand™
STE50VAR |
50 L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
STE82VMB
|
Fisherbrand™
STE82VMB |
82 L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Door Mechanism: Hand wheel type of quick-open door with safety lock system
- Control System: Micro-computer controlled with LCD display and touch-type keys
- Independent Temperature and Pressure Monitoring: Ensures precise control during sterilization processes
- High-Precision PT100 Sensor: Accurately controls liquid temperature
- Built-in Steam Generator: Provides efficient steam production for sterilization
- Overheating and Pressure Overload Protection: Enhances safety during operation
Specifications
Autoclavable | |
Decontamination of Biohazardous Waste | |
249°F to 275°F | |
Stainless Steel | |
Sterile |
Quality Management System | |
Autoclave | |
121°C to 135°C | |
Yes |