missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, Preserved with Salicylic Acid (Mercury Free), Certified, 1%, LabChem™
$38.43 - $468.00
Specifications
Chemical Name or Material | Starch Indicator |
---|---|
Name Note | Preserved with Salicylic Acid (Mercury Free) |
CAS Max % | 98.9,1,0.1 |
CAS | 9005-25-8,69-72-7 |
Grade | Certified |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
LC253101
|
LabChem
LC253101 |
500 mL | Poly Bottle |
Each for $38.43
|
|
||||
LC253102
|
LabChem
LC253102 |
1 L | Poly Bottle |
Each for $49.87
|
|
||||
LC253104
|
LabChem
LC253104 |
4 L | Poly Bottle |
Each for $108.73
|
|
||||
LC253106
|
LabChem
LC253106 |
10 L | Cubitainer™ |
Each for $300.87
|
|
||||
LC253105
|
LabChem
LC253105 |
20 L | Cubitainer™ |
Each for $468.00
|
|
||||
Specifications
Starch Indicator | |
98.9,1,0.1 | |
Certified |
Preserved with Salicylic Acid (Mercury Free) | |
9005-25-8,69-72-7 | |
Liquid |